Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339193.1 | NP_001326667.1 | 6 - 7 | 1305 | 89 | |
NM_114379.3 | NP_566874.1 | 6 - 7 | 1159 | 89 | ||
NM_180327.2 | NP_850658.1 | 6 - 7 | 1159 | 89 | ||
GenBank mRNA | AY039602.1 | - | 6 - 7 | 1156 | 89 | |
AY081726.1 | - | 5 - 6 | 1059 | 89 | ||
KJ138805.1 | - | 5 - 6 | 1059 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |