Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114613.3 | NP_190329.2 | 1 - 2 | 503 | 126 | |
NM_180335.1 | NP_850666.1 | 1 - 2 | 503 | 126 | ||
GenBank mRNA | AK226195.1 | - | 1 - 2 | 508 | 126 | |
BT015353.1 | - | 1 - 2 | 408 | 126 | ||
BT015887.1 | - | 1 - 2 | 405 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |