Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115261.4 | NP_566993.1 | 17 - 18 | 1633 | 68 | |
NM_180370.3 | NP_850701.1 | 16 - 17 | 1663 | 68 | ||
GenBank mRNA | AY056265.1 | - | 17 - 18 | 1565 | 68 | |
AY096635.1 | - | 17 - 18 | 1412 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |