Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084819.1 | NP_001078288.1 | 2 - 3 | 480 | 79 | |
NM_115346.4 | NP_191049.1 | 2 - 3 | 553 | 79 | ||
NM_180374.2 | NP_850705.1 | 2 - 3 | 480 | 79 | ||
NM_180375.2 | NP_850706.1 | 2 - 3 | 480 | 79 | ||
GenBank mRNA | AF325016.2 | - | 2 - 3 | 365 | 79 | |
AF326866.1 | - | 2 - 3 | 365 | 79 | ||
AF339688.1 | - | 2 - 3 | 299 | 79 | ||
AF361847.1 | - | 2 - 3 | 365 | 79 | ||
AY070473.1 | - | 2 - 3 | 365 | 79 | ||
AY094437.1 | - | 2 - 3 | 365 | 79 | ||
BT000852.1 | - | 2 - 3 | 299 | 79 | ||
X56062.1 | - | 406 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |