Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115860.4 | NP_191556.1 | 8 - 9 | 1508 | 154 | |
NM_180392.1 | NP_850723.1 | 7 - 8 | 1546 | 154 | ||
GenBank mRNA | AF181966.1 | - | 8 - 9 | 1431 | 154 | |
AY070034.1 | - | 8 - 9 | 1451 | 154 | ||
AY122922.1 | - | 8 - 9 | 1369 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |