Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035689.2 | NP_001030766.1 | 10 - 11 | 1111 | 116 | |
NM_001035690.1 | NP_001030767.1 | 11 - 12 | 1170 | 116 | ||
NM_001338758.1 | NP_001327446.1 | 10 - 11 | 1275 | 116 | ||
NM_001338759.1 | NP_001327445.1 | 9 - 10 | 720 | 116 | ||
NM_113456.4 | NP_189186.1 | 11 - 12 | 1174 | 116 | ||
NM_180413.2 | NP_850744.1 | 11 - 12 | 973 | 116 | ||
GenBank mRNA | AF091844.1 | - | 10 - 11 | 813 | 116 | |
AF360151.1 | - | 11 - 12 | 1038 | 116 | ||
AY054269.1 | - | 11 - 12 | 1022 | 116 | ||
AY142609.1 | - | 10 - 11 | 810 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |