Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035689.2 | NP_001030766.1 | 13 - 14 | 1326 | 92 | |
NM_001035690.1 | NP_001030767.1 | 14 - 15 | 1385 | 92 | ||
NM_001338758.1 | NP_001327446.1 | 13 - 14 | 1490 | 92 | ||
NM_001338759.1 | NP_001327445.1 | 12 - 13 | 935 | 92 | ||
NM_113456.4 | NP_189186.1 | 14 - 15 | 1389 | 92 | ||
NM_180413.2 | NP_850744.1 | 14 - 15 | 1188 | 92 | ||
GenBank mRNA | AF091844.1 | - | 13 - 14 | 1028 | 92 | |
AF360151.1 | - | 14 - 15 | 1253 | 92 | ||
AY054269.1 | - | 14 - 15 | 1237 | 92 | ||
AY142609.1 | - | 13 - 14 | 1025 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |