Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035689.2 | NP_001030766.1 | 6 - 7 | 874 | 115 | |
NM_001035690.1 | NP_001030767.1 | 7 - 8 | 933 | 115 | ||
NM_001338758.1 | NP_001327446.1 | 6 - 7 | 1038 | 115 | ||
NM_001338759.1 | NP_001327445.1 | 5 - 6 | 483 | 115 | ||
NM_113456.4 | NP_189186.1 | 7 - 8 | 937 | 115 | ||
NM_180413.2 | NP_850744.1 | 7 - 8 | 736 | 115 | ||
GenBank mRNA | AF091844.1 | - | 6 - 7 | 576 | 115 | |
AF360151.1 | - | 7 - 8 | 801 | 115 | ||
AY054269.1 | - | 7 - 8 | 785 | 115 | ||
AY142609.1 | - | 6 - 7 | 573 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |