Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120496.3 | NP_568134.1 | 8 - 9 | 1770 | 125 | |
NM_180432.2 | NP_850763.1 | 7 - 8 | 1602 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |