Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121867.4 | NP_568365.2 | 18 - 19 | 2531 | 111 | |
NM_180516.2 | NP_850847.1 | 18 - 19 | 2529 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |