Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198248.1 | NP_001185177.1 | 19 - 20 | 2221 | 90 | |
NM_001333364.1 | NP_001320175.1 | 19 - 20 | 2236 | 90 | ||
NM_103797.4 | NP_175333.3 | 19 - 20 | 2173 | 90 | ||
NM_180628.4 | NP_850959.1 | 19 - 20 | 2259 | 90 | ||
GenBank mRNA | AY082605.1 | - | 19 - 20 | 2066 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |