Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333397.1 | NP_001320454.1 | 4 - 5 | 780 | 117 | |
NM_103851.3 | NP_175386.2 | 4 - 5 | 809 | 117 | ||
NM_180630.2 | NP_850961.1 | 4 - 5 | 922 | 117 | ||
NM_180631.2 | NP_850962.1 | 4 - 5 | 922 | 117 | ||
GenBank mRNA | AY074305.1 | - | 4 - 5 | 809 | 117 | |
BT004376.1 | - | 4 - 5 | 764 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |