Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120506.5 | NP_196044.2 | 1 - 2 | 1447 | 131 | |
GenBank mRNA | AY664500.1 | - | 1 - 2 | 1056 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |