Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342778.1 | NP_001330248.1 | 2 - 3 | 583 | 79 | |
NM_120531.3 | NP_196069.1 | 2 - 3 | 658 | 79 | ||
GenBank mRNA | BT021123.1 | - | 2 - 3 | 386 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |