Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085058.2 | NP_001078527.1 | 4 - 5 | 3320 | 79 | |
NM_001342781.1 | NP_001331593.1 | 4 - 5 | 3325 | 79 | ||
NM_120538.2 | NP_196076.2 | 2 - 3 | 3735 | 79 | ||
GenBank mRNA | DQ335243.1 | - | 4 - 5 | 3232 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |