Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120541.4 | NP_196079.1 | 5 - 6 | 1930 | 87 | |
GenBank mRNA | AF325027.2 | - | 5 - 6 | 1644 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |