Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342782.1 | NP_001332170.1 | 7 - 8 | 1164 | 76 | |
NM_120544.3 | NP_196082.2 | 6 - 7 | 1397 | 76 | ||
NM_203002.4 | NP_974731.1 | 7 - 8 | 1186 | 76 | ||
GenBank mRNA | AK119095.1 | - | 6 - 7 | 1397 | 76 | |
BT026507.1 | - | 5 - 6 | 588 | 76 | ||
DQ017966.1 | - | 7 - 8 | 1005 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |