Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP77A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120548.3 | NP_196086.1 | 1 | 286 | 126 |
Gene Symbol: AT5G04670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120549.3 | NP_196087.1 | 4 | 4766 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |