Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120571.3 | NP_568144.1 | 1 - 2 | 211 | 72 | |
GenBank mRNA | AF208051.1 | - | 1 - 2 | 153 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |