Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125703.2 | NP_001119175.1 | 9 - 10 | 1084 | 127 | |
NM_120636.4 | NP_196173.1 | 9 - 10 | 1084 | 127 | ||
GenBank mRNA | AK118437.1 | - | 9 - 10 | 1060 | 127 | |
BT009685.1 | - | 9 - 10 | 947 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |