Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203302.1 | NP_001190231.1 | 4 - 5 | 817 | 82 | |
NM_120655.4 | NP_196192.1 | 4 - 5 | 840 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |