Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203303.1 | NP_001190232.1 | 5 - 6 | 695 | 124 | |
NM_120656.3 | NP_196193.1 | 4 - 5 | 693 | 124 | ||
NM_203007.3 | NP_974736.1 | 5 - 6 | 742 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |