Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120674.4 | NP_196211.1 | 2 - 3 | 526 | 154 | |
GenBank mRNA | AY087829.1 | - | 2 - 3 | 485 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |