Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342930.1 | NP_001331523.1 | 7 - 8 | 529 | 106 | |
NM_120800.4 | NP_196335.2 | 9 - 10 | 1084 | 106 | ||
GenBank mRNA | AY244746.1 | - | 9 - 10 | 914 | 106 | |
FJ708772.1 | - | 9 - 10 | 729 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |