Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342931.1 | NP_001331229.1 | 7 - 8 | 1741 | 121 | |
NM_120803.1 | NP_196338.1 | 7 - 8 | 1765 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |