Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120811.5 | NP_196346.1 | 10 - 11 | 2982 | 147 | |
GenBank mRNA | AK226960.1 | - | 10 - 11 | 2726 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |