Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120865.5 | NP_196400.2 | 2 - 3 | 623 | 123 | |
GenBank mRNA | BT015345.1 | - | 2 - 3 | 404 | 123 | |
BT015800.1 | - | 1 - 2 | 224 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |