Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127748.4 | NP_179770.1 | 4 - 5 | 909 | 97 | |
GenBank mRNA | AY035080.1 | - | 4 - 5 | 815 | 97 | |
BT008573.1 | - | 4 - 5 | 641 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |