Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124885.1 | NP_001118357.1 | 4 - 5 | 462 | 82 | |
NM_001335777.1 | NP_001323597.1 | 3 - 4 | 434 | 82 | ||
NM_001335778.1 | NP_001323598.1 | 4 - 5 | 334 | 82 | ||
NM_127757.3 | NP_565521.1 | 4 - 5 | 591 | 82 | ||
GenBank mRNA | AB071852.1 | - | 4 - 5 | 335 | 82 | |
AK229492.1 | - | 4 - 5 | 486 | 82 | ||
AY084960.1 | - | 4 - 5 | 444 | 82 | ||
BT024593.1 | - | 4 - 5 | 335 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |