Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127848.4 | NP_179868.1 | 5 - 6 | 1771 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |