Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335827.1 | NP_001325019.1 | 4 - 5 | 552 | 96 | |
NM_127857.5 | NP_179876.1 | 4 - 5 | 669 | 96 | ||
NM_179702.1 | NP_850033.1 | 4 - 5 | 472 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |