Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036324.1 | NP_001031401.1 | 6 - 7 | 638 | 102 | |
NM_001084476.1 | NP_001077945.1 | 6 - 7 | 638 | 102 | ||
NM_001335831.1 | NP_001323542.1 | 6 - 7 | 643 | 102 | ||
NM_127862.4 | NP_179880.1 | 6 - 7 | 742 | 102 | ||
GenBank mRNA | AY039596.1 | - | 6 - 7 | 631 | 102 | |
AY113074.1 | - | 6 - 7 | 616 | 102 | ||
BT000747.1 | - | 6 - 7 | 638 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |