Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161057.2 | NP_001154529.1 | 6 - 7 | 1078 | 123 | |
NM_127864.3 | NP_565546.3 | 6 - 7 | 724 | 123 | ||
NM_179703.2 | NP_850034.1 | 7 - 8 | 974 | 123 | ||
NM_179704.1 | NP_850035.1 | 7 - 8 | 778 | 123 | ||
NM_179705.1 | NP_850036.1 | 7 - 8 | 778 | 123 | ||
NM_201787.2 | NP_973516.2 | 7 - 8 | 751 | 123 | ||
GenBank mRNA | AF275313.1 | - | 7 - 8 | 123 | ||
AY035052.1 | - | 7 - 8 | 778 | 123 | ||
AY051060.1 | - | 7 - 8 | 701 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |