Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161057.2 | NP_001154529.1 | 9 - 10 | 1322 | 98 | |
NM_127864.3 | NP_565546.3 | 9 - 10 | 968 | 98 | ||
NM_179703.2 | NP_850034.1 | 10 - 11 | 1218 | 98 | ||
NM_179704.1 | NP_850035.1 | 10 - 11 | 1022 | 98 | ||
NM_179705.1 | NP_850036.1 | 10 - 11 | 1022 | 98 | ||
NM_201787.2 | NP_973516.2 | 10 - 11 | 995 | 98 | ||
GenBank mRNA | AF275313.1 | - | 10 - 11 | 1008 | 98 | |
AY035052.1 | - | 10 - 11 | 1022 | 98 | ||
AY051060.1 | - | 10 - 11 | 945 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |