Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335835.1 | NP_001325415.1 | 2 - 3 | 111 | 69 | |
NM_127869.3 | NP_179887.1 | 2 - 3 | 279 | 69 | ||
GenBank mRNA | AK117486.1 | - | 2 - 3 | 233 | 69 | |
AY102541.1 | - | 2 - 3 | 264 | 69 | ||
AY954795.1 | - | 2 - 3 | 136 | 69 | ||
BT005613.1 | - | 2 - 3 | 136 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |