Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335847.1 | NP_001324816.1 | 10 - 11 | 1788 | 97 | |
NM_127902.6 | NP_179919.1 | 15 - 16 | 2913 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |