Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127907.4 | NP_179924.1 | 1 - 2 | 371 | 88 | |
GenBank mRNA | BT028986.1 | - | 1 - 2 | 202 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |