Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202976.1 | NP_001189905.1 | 9 - 10 | 1318 | 101 | |
NM_001338190.1 | NP_001325883.1 | 8 - 9 | 1550 | 101 | ||
NM_112458.5 | NP_188209.3 | 9 - 10 | 1314 | 101 | ||
NM_180672.4 | NP_851003.2 | 9 - 10 | 1318 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |