Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114685.4 | NP_190399.2 | 3 - 4 | 559 | 121 | |
NM_180681.1 | NP_851012.1 | 3 - 4 | 559 | 121 | ||
GenBank mRNA | AB074533.1 | - | 3 - 4 | 249 | 121 | |
AJ417836.1 | - | 3 - 4 | 256 | 121 | ||
BT004258.1 | - | 3 - 4 | 559 | 121 | ||
BT005524.1 | - | 3 - 4 | 249 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |