Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122011.4 | NP_568390.2 | 7 - 8 | 1165 | 70 | |
NM_180712.3 | NP_851043.1 | 7 - 8 | 1165 | 70 | ||
GenBank mRNA | AB062615.1 | - | 7 - 8 | 975 | 70 | |
AF360208.1 | - | 7 - 8 | 1090 | 70 | ||
AY040056.1 | - | 7 - 8 | 975 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |