Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036833.2 | NP_001031910.1 | 1 - 2 | 389 | 89 | |
NM_001203418.1 | NP_001190347.1 | 2 - 3 | 296 | 89 | ||
NM_122032.6 | NP_197525.1 | 2 - 3 | 375 | 89 | ||
GenBank mRNA | AK227214.1 | - | 2 - 3 | 279 | 89 | |
AK316714.1 | - | 2 - 3 | 141 | 89 | ||
AY090237.1 | - | 2 - 3 | 282 | 89 | ||
BT000632.1 | - | 2 - 3 | 225 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |