Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122367.4 | NP_197847.3 | 5 | 1872 | 102 | |
GenBank mRNA | AF281062.1 | - | 5 | 1716 | 102 | |
BT002012.1 | - | 5 | 1729 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |