Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125941.1 | NP_001119413.1 | 4 - 5 | 677 | 72 | |
NM_124502.3 | NP_199936.2 | 5 - 6 | 669 | 72 | ||
NM_180837.3 | NP_851168.1 | 5 - 6 | 761 | 72 | ||
GenBank mRNA | AB053171.1 | - | 5 - 6 | 606 | 72 | |
AY140086.1 | - | 5 - 6 | 618 | 72 | ||
BT008388.1 | - | 4 - 5 | 358 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |