Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATTPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344937.1 | NP_001332431.1 | 10 - 11 | 1420 | 110 | |
NM_001344938.1 | NP_001332432.1 | 10 - 11 | 1313 | 110 | ||
NM_124525.2 | NP_199959.2 | 9 - 10 | 1184 | 110 | ||
NM_180840.2 | NP_851171.1 | 10 - 11 | 1424 | 110 | ||
NM_203193.3 | NP_974922.1 | 9 - 10 | 1737 | 110 | ||
GenBank mRNA | AY093147.1 | - | 9 - 10 | 1184 | 110 | |
BT008467.1 | - | 9 - 10 | 1005 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |