Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125995.2 | NP_001119467.1 | 10 - 11 | 1485 | 66 | |
NM_001345427.1 | NP_001332136.1 | 10 - 11 | 1676 | 66 | ||
NM_001345428.1 | NP_001332137.1 | 10 - 11 | 1572 | 66 | ||
NM_125453.6 | NP_200868.2 | 10 - 11 | 1401 | 66 | ||
NM_180902.5 | NP_851233.1 | 10 - 11 | 1485 | 66 | ||
GenBank mRNA | AF434673.1 | - | 10 - 11 | 1341 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |