Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203662.1 | NP_001190591.1 | 3 - 4 | 673 | 67 | |
NM_001345519.1 | NP_001330603.1 | 2 - 3 | 775 | 67 | ||
NM_125593.5 | NP_201006.2 | 3 - 4 | 840 | 67 | ||
NM_180913.4 | NP_851244.1 | 2 - 3 | 924 | 67 | ||
NM_203251.4 | NP_974980.1 | 3 - 4 | 679 | 67 | ||
GenBank mRNA | AK221282.1 | - | 3 - 4 | 562 | 67 | |
AK221305.1 | - | 3 - 4 | 583 | 67 | ||
AY669787.1 | - | 2 - 3 | 274 | 67 | ||
BT000784.1 | - | 2 - 3 | 775 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |