Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125784.3 | NP_568981.2 | 7 - 8 | 684 | 97 | |
NM_180929.4 | NP_851260.1 | 6 - 7 | 711 | 97 | ||
GenBank mRNA | AB027709.1 | - | 7 - 8 | 631 | 97 | |
AY039881.1 | - | 7 - 8 | 673 | 97 | ||
AY143900.1 | - | 7 - 8 | 582 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |