Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037070.1 | NP_001032147.1 | 7 - 8 | 945 | 118 | |
NM_001037071.1 | NP_001032148.1 | 7 - 8 | 852 | 118 | ||
NM_001037072.2 | NP_001032149.1 | 8 - 9 | 872 | 118 | ||
NM_001085331.1 | NP_001078800.1 | 7 - 8 | 1161 | 118 | ||
NM_001345675.1 | NP_001318881.1 | 7 - 8 | 1182 | 118 | ||
NM_125919.3 | NP_201324.1 | 6 - 7 | 1642 | 118 | ||
NM_180942.3 | NP_851273.1 | 7 - 8 | 1255 | 118 | ||
GenBank mRNA | AK317142.1 | - | 7 - 8 | 853 | 118 | |
AK317488.1 | - | 8 - 9 | 873 | 118 | ||
AY099593.1 | - | 7 - 8 | 971 | 118 | ||
AY128838.1 | - | 6 - 7 | 723 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |