Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203135.1 | NP_001190064.1 | 1 - 2 | 172 | 85 | |
NM_115068.5 | NP_190777.1 | 1 - 2 | 149 | 85 | ||
GenBank mRNA | BT005984.1 | - | 1 - 2 | 47 | 85 | |
U28048.1 | - | 1 - 2 | 98 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |