Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203135.1 | NP_001190064.1 | 2 - 3 | 270 | 81 | |
NM_115068.5 | NP_190777.1 | 2 - 3 | 247 | 81 | ||
GenBank mRNA | BT005984.1 | - | 2 - 3 | 145 | 81 | |
U28048.1 | - | 2 - 3 | 196 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |